Anti-Neuron specific enolase Rabbit Polyclonal Antibody
Supplier: Diagnostic Biosystems
RP054-01 RP054-05 RP054
76635-072EA 108.26 USD
76635-072 76635-074 76635-076
Anti-Neuron specific enolase Rabbit Polyclonal Antibody
Antibodies
This antibody is specific to γ-γ enolase isozyme of 46 kDa. This antibody stains neurons, neuroendocrine cells, gangliomas, oat cell carcinomas, astrocytomas, paragangliomas, glioblastomas, pheochromocytomas, melanomas and schwannomas.