Anti-Mesothelian Rabbit Monoclonal Antibody [clone: YP158]
Supplier: Diagnostic Biosystems
RMAB107-01 RMAB107-05 RMAB107
76634-820EA 114.63 USD
76634-820 76634-822 76634-824
Anti-Mesothelian Rabbit Monoclonal Antibody [clone: YP158]
Antibodies
Mesothelin is a glycocylphosphotidylianositol-linked cell-surface glycoprotein, which is present on the surface of normal mesothelium and is overexpressed in many patients with epithelial ovarian cancer and malignant mesotheliomas.