Anti-Adrenocorticotropic Hormone AH26 Mouse Monoclonal Antibody [clone: AH26]
Supplier: Diagnostic Biosystems
MOB244-01 MOB244-05 MOB244
76633-510EA 108.26 USD
76633-510 76633-512 76633-514
Anti-Adrenocorticotropic Hormone AH26 Mouse Monoclonal Antibody [clone: AH26]
Antibodies
This antibody is specific to an epitope on the N-terminus of ACTH. This antibody stains corticotrophs in the adenohypophysis and is useful in the classification of pituitary adenomas.