Anti-Collagen IV Mouse Monoclonal Antibody [clone: COL-94]
Supplier: Diagnostic Biosystems
MOB229-01 MOB229-05 MOB229
76633-474EA 108.26 USD
76633-474 76633-476 76633-478
Anti-Collagen IV Mouse Monoclonal Antibody [clone: COL-94]
Antibodies
This antibody recognizes collagen type IV epitope located on the α1 and/or α2 chains of human collagen type IV. This antibody shows no cross-reactivity with collagen types I, II, III, V, VI and VII and does not react with human vitronectin, fibronectin or chondroitin sulfate A, B and C.