Anti-Alpha Fetoprotein Mouse Monoclonal Antibody [clone: C3]
Supplier: Diagnostic Biosystems
MOB129-01 MOB129-05 MOB129
76633-352EA 108.26 USD
76633-352 76633-354 76633-356
Anti-Alpha Fetoprotein Mouse Monoclonal Antibody [clone: C3]
Antibodies
Alpha Fetoprotein (AFP) is a single chain glycoprotein of 70 kDa that is normally expressed in the fetal liver, gastrointestinal tract and yolk sac. This antibody recognizes an epitope of α-fetoprotein present in human and several other species.