Anti-Glycophorin C Mouse Monoclonal Antibody [clone: Ret40f]
Supplier: Diagnostic Biosystems
MOB067-01 MOB067-05 MOB067
76633-212EA 157.94 USD
76633-212 76633-214 76633-216
Anti-Glycophorin C Mouse Monoclonal Antibody [clone: Ret40f]
Antibodies
This antibody is specific to β sialoglycophorin also known as glycophorin C. Glycophorin C is found on the human erythrocyte membrane and is one of four sialic acid-rich polypeptides.