VWR® TAQ DNA Polymerase Master Mix
Supplier: VWR International
76620-456 76620-458 76620-460 76620-462 76620-450 76620-452 76620-454
76620-456EA 206.19 USD
76620-456 76620-458 76620-460 76620-462 76620-450 76620-452 76620-454
VWR® TAQ DNA Polymerase Master Mix
Nucleic Acid Reagents End-point PCR Enzymes and Kits
VWR® Taq DNA Polymerase Master Mix is a ready to use 1.1X or 2X reaction mix. Simply add primers, template and water to carry out primer extensions and other molecular biology applications.
  • All-in-one master mix for great convenience
  • Minimal optimization
  • Fewer pipetting steps
  • Increased reproducibility
  • High product yield
  • dUTP incorporation possible
  • Processes up to 5 kb
  • Leaves a 3'dA overhang
Taq DNA Polymerase master mixes are a good alternative to Taq DNA Polymerase.
The Taq DNA Polymerase master mixes offer reduced set-up time and eliminate the risk of contamination of reaction components. Fewer handling steps lead to better reproducibility, which makes master mixes suitable for standard PCR applications, screening and high-throughput testing.
Long term storage at -20 °C. Product expiry at -20 °C is stated on the label. Optional: Store at +4 °C for up to 6 months.
Available as 1.1X or 2X master mixes with a final MgCl2 concentration of either 1.5 mM or 2 mM.
Caution: For Research Use Only. Not for use in diagnostic procedures.
Order Now
SPECIFICATIONS
Environmentally Preferable
Application
Learn more