SteriKit® Sterile Vials, Stoppers, and Seals, Prince Sterilization Services
Supplier: Prince Sterilization Services
009 018 010 011 012 022 023 024 025 036 037 067 075 132
75805-150CS 10307.59 USD
75805-150 75805-152 75805-154 75805-156 75805-158 75805-160 75805-162 75805-164 75805-166 75805-168 75805-170 75805-172 75805-174 75805-176 75805-178 76614-780 76614-774 76614-782 76614-776 76614-784 76614-778 76614-768 76614-772 76614-770
SteriKit® Sterile Vials, Stoppers, and Seals, Prince Sterilization Services
Vials
The SteriKit® consists of ready to use vials, seals, and stoppers.
The vials are type I borosilicate glass and are USP WFI washed, aseptically repackaged in a 316 stainless steel tray, and USP dry heat depyrogenated in an ISO 7 cGMP cleanroom. The stoppers and seals are USP WFI washed, aseptically repackaged, and cGMP moist heat steam sterilized. Following sterilization, all components are assembled, vacuum packed, and heat sealed in a pre-sterilized LDPE bag under ISO 5 conditions.
All components are certified in a cGMP laboratories to be USP 85 pyrogen-free, USP 788 particulate-freeMore Product Information