Series 2000 Benchtop Meters, Environmental Express
Supplier: Antyila Scientific
35419-54 35419-55 35419-60 35419-62 35419-65 35419-67 35419-70 35419-72
76595-366EA 945.67 USD
76595-366 76595-368 76595-370 76595-372 76595-374 76595-376 76595-378 76595-380
Series 2000 Benchtop Meters, Environmental Express
Electrochemistry Meters pH and Multi-Parameter Meters
Developed using extensive feedback from laboratory staff, our new Environmental Express 2000 series meters are assembled in various configurations for measuring a pH/mV, EC, Ion, DO, BOD parameters.
  • Various configurations available to meet the demands of measuring pH/mV, EC, Ion, PC, PD, DO, BOD parameters
  • Automatic record retention and accessibility for continuous compliance with regulatory requirements
  • 0.01, 0.001 resolution for routine or research applications where acute accuracy is critical
  • Large displays with a small footprint, ideal for labs with
More Product Information