Qlicksmart Mounting Bracket for BladeFlask Removers, Cincinnati Surgical
Supplier: CINCINNATI SURGICAL CO.
48QSB
76583-022EA 18.77 USD
76583-022
Qlicksmart Mounting Bracket for BladeFlask Removers, Cincinnati Surgical
Blade Removers and Dispensers
This is the Universal Mounting Bracket used for the Qlicksmart BladeFlask and BladeFlask EVO scalpel blade removers to allow for single-handed blade removal operation.
  • Can be mounted on a wall, or on a trolley or bench
The mounting bracket is affixed using screws, or by using the extra strong double-sided adhesive on the bottom if screws cannot be used.