Swann Morton Surgical Blade Handle #8, Stainless Steel, Cincinnati Surgical
Supplier: CINCINNATI SURGICAL CO.
0780SM-S
76583-012EA 142.96 USD
76583-012
Swann Morton Surgical Blade Handle #8, Stainless Steel, Cincinnati Surgical
Scalpels and Scalpel Assemblies Scalpel Handles
Fits size 60 and 70 blades.
  • Autoclavable
This product is manufactured to ISO 13485 standards.