Biohazard Bags, Autoclavable, Mopec
Supplier: Mortech Manufacturing
BG680-C BG680-R
76579-390CS 387.25 USD
76579-390 76579-392
Biohazard Bags, Autoclavable, Mopec
Bags Waste Disposal Bags
Mopec’s autoclavable biohazard bags are specifically designed with seamless bottoms for durability and leak-resistance, and are made from a heavy-duty, puncture-resistant polypropylene. These bags can also withstand temperatures up to 250 ˚F during autoclaving. Each bag features the OSHA-standard 'Biohazard' symbol and words 'Hazardous Waste'.
  • Dimensions: 26L×12.2"W
  • Made from a heavy-duty, puncture-resistant polypropylene
  • Can withstand temperatures up to 250 ˚F during autoclaving
  • Feature OSHA-standard 'Biohazard' symbol and words 'Hazardous Waste'
  • Feature
More Product Information