Diyttrium trioxide 99,9%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Yttrium oxide, Yttrium (III) oxide

IN3862-25G
76561-066EA 21.81 USD
76561-066
Diyttrium trioxide 99,9%
Diyttrium trioxide
Formula: Y₂O₃
Boiling Pt: 4300 °C
Melting Pt: 2410 °C
Density: 4.84 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011473
CAS Number: 1314-36-9
EINECS: 215-233-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR