QUICK CHECK* Sterilization Wraps, Halyard
Supplier: O&M HALYARD, INC
34154 34170 34156 34192 34178 34176 34194 34200 34188 34161 34172 34167 34193 34174 34175 34185 34186 34149 34187 34168 34171 34173 34160 34189 34159 34181 34182 34190 37045 34183 34158 34184 34157 34145 34146 34191 34180 34162 34165 34199 34198 34148 34143 34195 34196 34179 34151 34163 34164 34147 34142 34197
76557-876CS 75.12 USD
76557-876 76557-878 76557-880 76557-882 76557-884 76557-886 76557-888 76557-890 76557-892 76557-894 76557-896 76557-898 76557-900 76557-902 76557-904 76557-906 76557-908 76557-910 76557-912 76557-914 76557-916 76557-918 76557-920 76557-922 76557-924 76557-926 76557-928 76557-930 76557-932 76557-934 76557-936 76557-938 76557-940 76557-942 76557-944 76557-946 76557-948 76557-950 76557-952 76557-954 76557-956 76557-958 76557-960 76557-962 76557-964 76557-966 76557-968 76557-970 76557-972 76557-974 76557-976 76557-978
QUICK CHECK* Sterilization Wraps, Halyard
Sterilization Wraps
Halyard QUICK CHECK sterilization wraps are the fast, easy way to ensure instrument sterility is uncompromised.
  • Two distinct, contrasting layers, translucent and opaque are bonded together making inspecting trays easier and faster than ever
  • Superior microbial barrier protection with Powerguard* technology
  • Cleared for all of the same sterilization systems as Halyard ONE-STEP Wrap
  • Made from recyclable #5 polypropylene fabric
To make specific trays stand out from the rest, simply flip the sheet and wrap with the other side out. Same protection, noMore Product Information