VWR® C-Leg Lab Benches, Frames with Notches for Uprights, Snow White
Supplier: VWR International
76550-302 76550-768 76551-022 76550-638 76550-702 76550-610 76551-132 76550-682 76550-496 76550-300 76550-430 76550-580 76551-048 76550-424 76550-844 76551-088 76550-408 76551-032 76550-388 76550-750 76550-562 76550-422 76550-756 76550-554 76550-386 76550-444 76551-026 76550-984 76550-680 76551-030 76551-104 76550-548 76550-928 76551-110 76550-824 76550-746 76550-512 76550-538 76550-550 76550-982 76550-716 76551-174 76550-338 76551-114 76550-644 76551-116 76550-850 76550-974 76550-888 76550-648 76550-704 76551-068 76550-394 76550-706 76550-858 76550-560 76550-646 76550-568 76550-940 76551-010
76550-302EA 4213.65 USD
76550-302 76550-768 76551-022 76550-638 76550-702 76550-610 76551-132 76550-682 76550-496 76550-300 76550-430 76550-580 76551-048 76550-424 76550-844 76551-088 76550-408 76551-032 76550-388 76550-750 76550-562 76550-422 76550-756 76550-554 76550-386 76550-444 76551-026 76550-984 76550-680 76551-030 76551-104 76550-548 76550-928 76551-110 76550-824 76550-746 76550-512 76550-538 76550-550 76550-982 76550-716 76551-174 76550-338 76551-114 76550-644 76551-116 76550-850 76550-974 76550-888 76550-648 76550-704 76551-068 76550-394 76550-706 76550-858 76550-560 76550-646 76550-568 76550-940 76551-010
VWR® C-Leg Lab Benches, Frames with Notches for Uprights, Snow White
Furniture Tables and Workbenches
VWR® lab benches are ergonomically designed to meet the needs of laboratory personnel. There are many possible adjustments, all components fit together seamlessly, and each detail is designed for real world functionality.
  • Snow white frame
  • Ergonomic and highly adaptable to the needs of the user
  • Flexible, modular design
  • Offering a wide range of accessories to meet any need
  • Various concept workbench sizes and combinations
  • Height adjustment available either electrically (electric motor height adjustment) or with a hand crank or an Allen key
  • A wide range of
More Product Information