VWR® Digital Dry Bath Block Heaters
Supplier: VWR International
BSH1001-VWR BSH1002-VWR BSH1004-VWR
76549-936EA 796.57 USD
76549-936 76549-938 76549-940
VWR® Digital Dry Bath Block Heaters
Block Heaters Digital Block Heaters
VWR® digital dry bath block heaters, for one, two or four blocks. For accurate and uniform incubation of tubes and microplates. Accurately controls temperaure up to 150 °C.
  • Digital temperature control
  • Wide range of block accessories for all common tubes and plates
  • Available in single, dual and four block capacity
Certifications: CE marked.
Ordering information: Heaters require a VWR® modular heating block for operation (sold separately).
Order Now
SPECIFICATIONS
Environmentally Preferable
Timer Range 1 to 999 min. or ON
Increments 0.1 °C