VWR® Magnetic Hotplate Stirrers
Supplier: VWR International
76549-912 76549-918 76549-920 76549-926
76549-912EA 550.6 USD
76549-912 76549-918 76549-920 76549-926
VWR® Magnetic Hotplate Stirrers
Stirrers Magnetic Hotplate Stirrers
VWR® magnetic hotplate stirrers, with temperature control up to 400 °C and stirring speeds up to 1500 rpm.
  • White ceramic surface for excellent chemical resistance
  • Digital speed control up to 400 °C
  • Available with external temperature probe (displays actual sample temperature)
  • 'HOT' LED safety indicator alerts user when plate is unsafe to touch
Corresponding magnetic stirrers (stirring only 76549-916 and 76549-924) or hotplates (heating only 76549-914 and76549-922) with ceramic plates are also available. Full specification details are given underMore Product Information