Scalpel Handle, Stainless Steel, No. 4L, Mortech
Supplier: Mortech Manufacturing
AJ102
76549-386EA 55.41 USD
76549-386
Scalpel Handle, Stainless Steel, No. 4L, Mortech
Scalpels and Scalpel Assemblies Scalpel Handles
Stainless steel scalpel blade handle with extended grip.
  • Premium grade stainless steel
  • Ridged grip grasp
  • Fits #20, #21, #22 and #23 scalpel blades
Works for #20 thru #23 scalpel blades. Compatible with QlickSmart Blade Removal System. Durable and reusable.