Knife, Large Grossing, Mortech
Supplier: Mortech Manufacturing
AH110
76549-364EA 258.86 USD
76549-364
Knife, Large Grossing, Mortech
Knives
Sani-Safe large autopsy grossing knife has a non-slip safety handle that comfortably molds to your hand reducing fatigue. Handle is securely bonded to the cutting blade and the honed cutting edge makes for the perfect autopsy knife.
  • Premium grade stainless steel with plastic handle
  • Honed cutting edge with air-gap grooves
Well-balanced blade is hand ground and honed and can be constantly sharpened for long life. Easily sanitized with any disinfectant.