Scissors, Titanium, Iris, Mortech
Supplier: Mortech Manufacturing
AA707 AA708
76548-622EA 215.71 USD
76548-622 76548-624
Scissors, Titanium, Iris, Mortech
Scissors
Titanium scissors weigh approximately 25% less than stainless steel, making them the first choice for many technicians. Titanium alloy instruments keep edges sharp longer than traditional instruments. More stain resistant than stainless steel. Helpful in cutting into the facial region.
  • Standard grade titanium stainless steel
Available with straight or curved edges. Do not use bleach or other caustic chemicals.