Fixative Dissect Aid™, Statlab
Supplier: StatLab
9001-1 9001-2.5 9001-5
76548-062EA 73.86 USD
76548-062 76548-064 76548-066
Fixative Dissect Aid™, Statlab
Stains Microscopy and staining reagents
Dissect-aid is a fixative which helps make lymph nodes more identifiable during pathology examination.
  • The product turns lymph nodes pale/white so they stand out from other tissue
  • Solution firms the nodes for touch identification when performing gross examination
  • Dramatically improves lymph node recovery/harvesting, and also an excellent fixative for fatty tissues
  • This formulation has helped in diagnosis for the past 30 years
Part of the StatLab line of prefilled bulk containers.
Manufactured in the USA