Virex® II 256 One-Step Disinfectant Cleaner and Deodorant
Supplier: DIVERSEY INC.
04332 5019317
76547-674CT 212.25 USD
76547-674 76547-694
Virex® II 256 One-Step Disinfectant Cleaner and Deodorant
Cleaning Agents Disinfectants
A one-step, quaternary-based disinfectant cleaner concentrate providing broad spectrum disinfection at 1:256 dilution.
  • Kills microorganisms including HIV-1, VRE, MRSA, GRSA, MRSE, VISA, PRSP, Herpes Simplex Types 1&2, Influenza Type A2, Adenovirus, Rotavirus and many more
  • Kills the organisms that cause odors and works as an odor counteractant to further eliminate odors not associated with bacteria.
  • Highly concentrated quaternary formula provides excellent one-step, cost-effective cleaning and disinfection
Use in healthcare and other facilitiesMore Product Information