Cole-Parmer® Large Capacity Analog Infrared Hotplate, Antylia Scientific
Supplier: Antyila Scientific
04807-85
76539-138EA 1447.12 USD
76539-138
Cole-Parmer® Large Capacity Analog Infrared Hotplate, Antylia Scientific
Heaters Hot Plates
Easy to use temperature control dial.
  • Infrared heater is efficient and saves time and energy
  • Ideal for heating large volumes of liquid
  • Chemically resistant ceramic top
Hotplate temperature is controlled by an easy-to-use dial. Each model features a ‘Hot’ warning light which flashes whenever the plate temperature is above 50 °C.
Order Now
SPECIFICATIONS
Environmentally Preferable