Muffle Furnaces up to 1200 °C, Models L and LT, Nabertherm
Supplier: NABERTHERM INC.
L-014K27N1-120 L-014K27N1-240 L-034H2LN2-120 L-034H2LN2-208 L-034H2LN2-240 L-034H2ON2-120 L-034H2ON2-208 L-034H2ON2-240 L-034K2LN2-120 L-034K2LN2-208 L-034K2LN2-240 L-034K2ON2-120 L-034K2ON2-208 L-034K2ON2-240 L-054H2LN2-208 L-054H2LN2-240 L-054H2ON2-120 L-054H2ON2-208 L-054H2ON2-240 L-054K2LN2-208 L-054K2LN2-240 L-054K2ON2-208 L-054K2ON2-240 L-094H2ON2-208 L-094H2ON2-240 L-094K2LN2-208 L-094K2LN2-240 L-094K2ON2-208 L-094K2ON2-240 L-154H2LN2-208 L-154H2LN2-240 L-154H2ON2-208 L-154H2ON2-240 L-154K2LN2-208 L-154K2LN2-240 L-154K2ON2-208 L-154K2ON2-240 L-244H2LN2-480 L-244H2ON2-480 L-244K2LN2-480 L-244K2ON2-480 L-404H2LN2-480 L-404H2ON2-480 L-404K2LN2-480 L-404K2ON2-480
76537-634EA 3951 USD
76537-634 76537-636 76537-638 76537-640 76537-642 76537-644 76537-646 76537-648 76537-650 76537-652 76537-654 76537-656 76537-658 76537-660 76537-662 76537-664 76537-666 76537-668 76537-670 76537-672 76537-674 76537-676 76537-678 76537-680 76537-682 76537-684 76537-686 76537-688 76537-690 76537-692 76537-694 76537-696 76537-698 76537-700 76537-702 76537-704 76537-706 76537-708 76537-710 76537-712 76537-714 76537-716 76537-718 76537-720 76537-722 76537-724 76537-726 76537-728 76537-730 76537-732 76537-734
Muffle Furnaces up to 1200 °C, Models L and LT, Nabertherm
Ovens Furnaces (Ashing and Muffle Ovens)
These muffle furnaces have been proven for daily laboratory use. These models stand out for their workmanship, advanced and attractive design and high level of reliability. The muffle furnaces come equipped with either a fold down or lift up door at no extra charge.
  • Tmax 1200 °C
  • Heating from two sides by ceramic heating plates (heating from three sides for muffle furnaces L 24/11 - LT 40/12)
  • Thermocouple type S; manual or automatic gas supply system
  • Ceramic heating plates with integral heating element which is safeguarded and easy to replace
  • Optional fold
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable