Cole-Parmer® Essentials Carboy with Spigot, Rectangular, HDPE, Antylia Scientific
Supplier: Antyila Scientific
06066-30 06066-31
76534-252EA 195.07 USD
76534-252 76534-254
Cole-Parmer® Essentials Carboy with Spigot, Rectangular, HDPE, Antylia Scientific
Bottles Carboys
Don’t waste valuable bench space.
  • Leakproof for safe transport
These leakproof HDPE rectangular carboys with spigot easily store and dispense sterile water and other solutions. Space-saving design requires less space than traditional round carboys. Features include built-in stainless steel handle and wide mouth opening for easy cleaning. Carboys are not autoclavable.
Certifications: USP Class VI certified