SevenDirect™ SD30 Conductivity Meter
Supplier: Mettler Toledo
30671551 30671562 30671563
76533-430EA 1851.53 USD
76533-430 76533-452 76533-454
SevenDirect™ SD30 Conductivity Meter
Electrochemistry Meters Conductivity Meters
The conductivity meter SevenDirect™ SD30 provides accurate conductivity measurements for almost any sample across a wide range of applications. The EasyPlace™ electrode arm and protective cover are included with the instrument. The benchtop meter's ease of use, data handling options and robustness make it ideal for all industries.
  • Intuitive user interface using 7" touchscreen
  • On-screen instructions guide through the calibration process
  • EasyPlace™ electrode arm ensures precise placement of the sensor
  • Feature a closed connector compartment to prevent
More Product Information