SevenDirect™ SD20 pH Meter
Supplier: Mettler Toledo
30671550 30671554 30671556 30671557 30671558 30671559 30671560 30671561 30671555
76533-428EA 1318.75 USD
76533-428 76533-436 76533-438 76533-440 76533-442 76533-444 76533-446 76533-448 76533-450
SevenDirect™ SD20 pH Meter
Electrochemistry Meters pH and Multi-Parameter Meters
The pH meter SevenDirect™ SD20 provides accurate pH measurements for almost any sample across a wide range of applications. The EasyPlace™ electrode arm and protective cover are included with the instrument. The benchtop meter's ease of use, data handling options and robustness make it ideal for all industries.
  • Intuitive user interface using 7" touchscreen
  • On-screen instructions guide through the calibration process
  • EasyPlace™ electrode arm ensures precise placement of the sensor
  • Feature a closed connector compartment to prevent corrosion
  • A replacable
More Product Information