arium® comfort Pre-treatment Cartridge, Sartorius
Supplier: Sartorius
H2O-CPFCO-1
76533-172PK 624.65 USD
76533-172
arium® comfort Pre-treatment Cartridge, Sartorius
Water Purification Systems Pure and Ultra Pure Water Systems
Pre-filter consisting of high efficient activated carbon combined with a process optimizing catalyst. Removes reliable oxidizing agents, heavy metals, and particular impurities.
  • Fast and effective adsorption of impurities through high-grade activated carbon
  • Highly efficient catalyst for removing free chlorine
The combination of spherical, catalytically active activated carbon with an added catalyst constitutes reliable protection for a downstream reverse osmosis (RO) membrane. The cartridge removes oxidation agents, such as free chlorine and ozone,More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable
W×D×H 280×210×140 mm