Cole-Parmer® BM-400/ BM-450 Mixer/Mill® High-Energy Ball Mills
Supplier: SPEX
3111 3116 3116PC 8000M-230 8002 6133 6134 6135 8000D-230 8000M-115 8010 8011 8012 8000D-115 8015 8016 8017 6133PC-T 8018 8019
76181-852BG 70.86 USD
76181-852 76181-862 76181-864 76182-066 76182-072 76182-214 76182-218 76182-220 76182-242 76182-244 76182-392 76182-394 76182-396 76182-462 76182-516 76182-518 76182-520 76533-086 76533-148 76533-150
Cole-Parmer® BM-400/ BM-450 Mixer/Mill® High-Energy Ball Mills
Mills
Easily pulverize hard or brittle samples to analytical fineness in minutes.
  • Grind rocks, minerals, grains, pharmaceuticals, plant and animal tissue, cement, and ceramics
  • Accepts a wide selection of grinding and mixing vials
  • Programmable 100-minute timer
  • Safety interlock stops the motor if the lid is opened during operation
Use these powerful ball mills for mechanical alloying, slurry grinding, blending powders, mixing emulsions, and mechanochemistry. The sample is placed in a grinding vial with one or more balls, then shaken vigorously. The clampMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable