Erlenmeyer Flask Set of 5, Borosilicate Glass, United Scientific Supplies
Supplier: United Scientific Supplies
FGSET5
76531-944EA 36.87 USD
76531-944
Erlenmeyer Flask Set of 5, Borosilicate Glass, United Scientific Supplies
Flasks Erlenmeyer Flasks
Set of 5 narrow-mouth Erlenmeyer flasks.
  • Made from borosilicate glass for an excellent combination of thermal shock resistance and physical strength
  • Heavy-duty rims and durable white enamel graduations
A convenient set of five of narrow-mouth borosilicate glass Erlenmeyer flasks. Set consists of one each of 50 ml, 125 ml, 250 ml, 500 ml, and 1000 ml flasks.