Globe Glass™ Graduated Cylinders, To Deliver, Globe Scientific
Supplier: Globe Scientific
8300005 8300010 8300025 8300050 8300100 8300250 8300500 8301000 8302000 8330005 8330010 8330025 8330050 8330100 8330250 8330500 8331000 8332000
76523-316CS 131.56 USD
76523-316 76523-318 76523-320 76523-322 76523-324 76523-326 76523-328 76523-330 76523-332 76523-334 76523-336 76523-338 76523-340 76523-342 76523-344 76523-346 76523-348 76523-350
Globe Glass™ Graduated Cylinders, To Deliver, Globe Scientific
Cylinders Measuring Cylinders
Globe Glass™ graduated cylinders are calibrated to ASTM E1272 standards for precise measurement of volumes of liquids. A solid hexagonal base provides stability, and an included polyethylene (PE) bumper guard helps to protect against accidental breakage. These graduated cylinders are calibrated TD, 'To Deliver', and are available in sizes ranging from 5 to 2000 ml, with Class A or Class B calibration tolerance options. Class A graduated cylinders include a batch calibration certificate.
  • Calibrated TD; 'To Deliver'
  • Class A calibration tolerance for highest
More Product Information