Allegra V-15R Benchtop Centrifuges, Beckman Coulter
Supplier: Beckman Coulter
C63124 C63125 C63375 C63380 C63381 C63386 C63387 C63420 C63421 C63426 C63427 C63432 C63433 C63438 C63439 C63444 C63445 C63450 C69866 C69867
76521-700EA 12708.8 USD
76521-700 76521-702 76521-704 76521-706 76521-708 76521-710 76521-712 76521-714 76521-716 76521-718 76521-720 76521-722 76521-724 76521-726 76521-728 76521-730 76521-732 76521-734 76628-204 76628-206
Allegra V-15R Benchtop Centrifuges, Beckman Coulter
Centrifuges Benchtop Centrifuges
The Allegra V-15R benchtop centrifuges bring more versatility to the laboratory bench. These compact, refrigerated centrifuges are ideal for evolving laboratories.
  • Free up precious benchtop space
  • Support multiple workflows with just one instrument
  • Advanced temperature management
  • Wide choice of centrifuge packages available to suit various applications
The Allegra V-15R centrifuges maintain a consistent temperature, allowing you to spin faster and longer without overheating your samples. All while Rapid Temp and Eco Mode allow for even more efficiencyMore Product Information