Reagent Reservoirs, Irish Life Sciences
Supplier: Irish Life Sciences
RES8-350 RES12-351 RES96-352 RES96-352S RES384-353
76509-434CS 336.27 USD
76509-434 76509-436 76509-438 76509-440 76509-442
Reagent Reservoirs, Irish Life Sciences
Reagent Reservoirs
These reagent reservoirs were designed especially for use with automated liquid handling platforms. They can also be used with manual systems.
  • DNase/RNase free
  • Polymer selection to overcome the surface tension of the liquid and minimize residual liquid
  • Suitable for storage down to –80 °C
This reservoir has been developed precisely to conform to the footprint dimensions defined by SLAS/ANSI standards. This guarantees its compatibility with both manual and automated sample handling systems, readers, and microplate washers.
Certifications: More Product Information