Filtering Flasks, with Plastic Hose Connection, Sartorius
Supplier: Sartorius
166721
76500-764EA 511.49 USD
76500-764
Filtering Flasks, with Plastic Hose Connection, Sartorius
Flasks Filter Flasks
These vacuum-resistant filtering flasks made of DURAN® borosilicate glass, can be used in filtrate collection.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Standard according to DIN 12476.
Order Now
SPECIFICATIONS
Environmentally Preferable