Misty® Secure Hydrochloric Acid Bowl Cleaner, Mint Scent, Essendant
Supplier: Janitorial Supplies
AMR1038801
76500-092CT 88.83 USD
76500-092
Misty® Secure Hydrochloric Acid Bowl Cleaner, Mint Scent, Essendant
Cleaning Agents Disinfectants
Fast acting 9% hydrochloric acid toilet bowl and urinal cleaner removes stains, hard water deposits, and organic matter with ease.
  • Keeps bowls and urinals bright, clean, and fresh smelling
  • 9% hydrochloric acid formula
  • Thickened formula clings to surfaces for prolonged contact time
  • Odor counteractants kills offensive odors
  • Non-fuming
A fast acting 9% hydrochloric acid toilet bowl and urinal cleaner is suitable for use on ceramics, enamel, or metal. Cleaner works against bacteria, grime, mildew, mineral scale, mold, organic matter, rust, soil, and stains.