Septihol® Hand Sanitizer
Supplier: Steris
625736
76499-224CS 757.98 USD
76499-224
Septihol® Hand Sanitizer
Skincare Skin Disinfectants Liquid and Foam Skin Disinfectants
Septihol® Hand Sanitizer is an alcohol-based hand rub that offers fast antimicrobial action on hands without the need for a water rinse.
  • Fast acting, proven antimicrobial efficacy for hands
  • Formulated with 62% (v/v) ethyl alcohol
  • Lightweight foam mousse format allows the best possible hand coverage
  • No rinse format with minimal residue on hands
  • Manufactured under a cGMP, FDA compliant quality system
  • Fragrance-free
Septihol® Hand Sanitizer is a luxurious foam sanitizer formulated with 62% v/v ethyl alcohol to provide a convenient method forMore Product Information