Mopec Bone Dust Vacuum, Mortech®
Supplier: Mortech Manufacturing
BD080
76498-346EA 7608.6 USD
76498-346
Mopec Bone Dust Vacuum, Mortech®
Vacuum Cleaners
HEPA filtration minimizes exposure to particulate matter. The bone dust vacuum collector attaches directly to the autopsy saw for safety and efficiency.
  • One-year warranty
  • 115 V, 60 Hz with US plug
  • Reduced noise level
  • HE-rated filter is powerful enough for even the smallest particles
  • Disposable filter cartridge is easy to replace
  • 10′ power cord
HEPA vacuum includes HEPA filter, vacuum hose (102"), and 'Y' bracket. The vacuum is designed specifically for use with Stryker 810 Autopsy Saw, Mopec 810 Autopsy Saw, and Mopec 1000 Autopsy Saw. Not for useMore Product Information