VWR® Environmentally Friendly Thermometers, Double Safe
Supplier: VWR International
60400-0111 60400-0611 60400-0711 60400-1211 60400-1811
76491-608CS 862.29 USD
76491-608 76491-610 76491-612 76491-614 76491-616
VWR® Environmentally Friendly Thermometers, Double Safe
Thermometers Analog Thermometers
These thermometers are easy to read quickly and accurately due to the high contrast black numbering against a yellow background. The numbers are printed larger than on a standard thermometer. Ideal for use in chemical/petrochemical laboratories, universities, scientific institutions, food and beverage processing, pharmaceutical and medical industries, dairies, breweries and distilleries.
  • Enviro safe liquid
  • Non-toxic, non-hazardous, biodegradable liquid
  • PFA coating prevents any contamination of liquid or glass
Includes a multi-language (English, French,More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable