Borosil® Volumetric Transfer Pipettes, Class B, Accuracy ASTM E-969-02, Bulb Pipette, Foxx Life Sciences®
Supplier: Foxx Life Sciences
7103001 7103002 7103003 7103004 7103005 7103006 7103007 7103008 7103009 7103010 7103011 7103012 7103016 7103706 7103707 7103708 7103709
76490-914CS 118.92 USD
76490-914 76490-916 76490-918 76490-920 76490-922 76490-924 76490-926 76490-928 76490-930 76490-932 76490-934 76490-936 76490-938 76490-940 76490-942 76490-944 76490-946
Borosil® Volumetric Transfer Pipettes, Class B, Accuracy ASTM E-969-02, Bulb Pipette, Foxx Life Sciences®
Pipettes Graduated Pipettes
Borosil® brand Pipettes are made of specially-selected, heavy wall, machine-drawn, borosilicate accurate, bore glass tubing.
  • Calibrated for delivery
  • Used for accurate measurement to delivery of fluids
  • Round
They have precision formed jets, and bevel ground ends that resist chipping. The graduations are made in durable white enamel that is resistant to laboratory reagents and repeat washing. All Pipettes are individually measured and then marked. The uniformity of bore ensures accuracy between any two readings and they are autoclavable. The Class BMore Product Information