ESD Rubber Table Mats, Low Profile, Transforming Technologies
Supplier: Transforming Technologies
MTLP2460B MTLP2460GY MTLP2460RB MTLP2472B MTLP2472GY MTLP2472RB MTLP3036B MTLP3036GY MTLP3036RB MTLP3048B MTLP3048GY MTLP3048RB MTLP3060B MTLP3060GY MTLP3060RB MTLP3072B MTLP3072GY MTLP3072RB MTLP3660B MTLP3660GY MTLP3660RB
76490-028EA 123.93 USD
76490-028 76490-030 76490-032 76490-034 76490-036 76490-038 76490-040 76490-042 76490-044 76490-046 76490-048 76490-050 76490-052 76490-054 76490-056 76490-058 76490-060 76490-062 76490-064 76490-066 76490-068
ESD Rubber Table Mats, Low Profile, Transforming Technologies
Surface Protectors Bench Protectors
The MT2500 series ESD rubber mats are made from proven 0.060" thick 2-ply rubber.
  • Dissipative work surface 10E6 to 10E8 ohms Rtt
  • Resistance to ground 8 × 10E6-8 ohms
  • Resistance point to point 2 × 10E6-8 ohms
A conductive layer backs the static dissipative top layer that creates outstanding electrical performance characteristics. The 2500 series offers excellent resistance to hot solders, soldering irons and most solvents. MT2500 series is available in 50' rolls in 24, 30, and 36" widths. Pre-cut mat sizes are packaged individually with two (2) female 1More Product Information