TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether) 10% in water, ReadiUse™ Peroxide-and Carbonyl-Free

Supplier: AAT BIOQUEST INC

Synonyms: Polyethyleneglycol-[4-(1,1,3,3-tetramethylbutyl)phenyl]-ether, (p-tert-Octylphenoxy)polyethoxyethanol, Octoxynol, Polyoxyethylene(10) Octylphenyl Ether, t-Octylphenoxypolyethoxyethanol, O-[4-(1,1,3,3-Tetramethylbutyl)phenoxy]polyethoxyethanol

60012
76485-144EA 124.31 USD
76485-144
TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether) 10% in water, ReadiUse™ Peroxide-and Carbonyl-Free
TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether)

Triton™ X-100 is a common non-ionic mild surfactant, which is often routinely utilized to lyse cells, extract proteins and cellular organelles, permeabilize the living cell membranes and solubilize proteins.


  • Ready to use
  • Free of all deleterious enzymes
  • Mild lysing agent


Caution: Research Use Only (RUO)

Melting Pt: 6
Flash Pt: 251
Storage Temperature: Ambient
MDL Number: MFCD00132505
CAS Number: 9002-93-1
Merck Index: 13,06793

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR