Differential Scanning Calorimetry, DSC 4000, PerkinElmer
Supplier: PerkinElmer
N5370213 N5370212
76474-848EA 46632.11 USD
76474-848 76474-850
Differential Scanning Calorimetry, DSC 4000, PerkinElmer
Calorimeters
The DSC 4000 provides dependable performance and quality results. This single-furnace, heat flux DSC is designed to run all your routine applications and includes an easy-to-load vertical furnace that is resistant to oxygen and chemical corrosion.
  • Integrated mass flow controller for convenient gas control and switching
  • Fast 45-position autosampler option for reliable unattended operation
  • Portable cooling device (PCD) for cost-effective operation at low temperature
  • Efficient, easy-to-clean nickel chromium sample plate
  • Temperature range: −100 to +450 °C
More Product Information