Alcohol Wipes, First Aid Only
Supplier: First Aid Only
12-011-002 FAE-4001-001 H305-001 H305-200-001
76048-930BX 2.3 USD
76048-930 76049-094 76049-192 76474-334
Alcohol Wipes, First Aid Only
Wipes Disposable Wipes and Towels Disinfectant Wipes
Prevent infections with single-use alcohol cleansing pads. Each pad is saturated with 70% isopropyl alcohol and helps kill the germs in the area surrounding the wound.
  • Cleanse and prevent infection in minor wounds
  • Single-use alcohol cleansing pads
Each wipe is individually packaged, and they do a great job of cleansing and preventing infection in minor cuts, scrapes, and abrasions.
The SmartCompliance wipes are compatible with cabinets: 90608, 90580, 90659, 90660, 746000, 746004, 746005, 746006, 90732, 90829, 90830, 90831, 90832, 90833, 90834 90835,More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable