Rapid Response™ Urinary Tract Infection Test Strip, BTNX

Supplier: BTNX INC.

UTI-1S1
76473-526KT 12.83 USD
76473-526
Rapid Response™ Urinary Tract Infection Test Strip, BTNX
Clinical Diagnostic Supplies Clinical Reagents and Kits

The Rapid Response® Urinary tract infection test strip is a rapid, self-performing test for the detection of leukocytes and nitrite in urine, to aid in the screening of urinary tract infection (UTI).


  • Sample: Urine
  • Format: Strip
  • Specificity: Leukocytes and nitrites
  • Accuracy: Leukocyte: 94.4%, Nitrite: 98.4%
  • Time to result: 1 to 2 minutes
  • Storage condition: 2 to 30 °C (36 to 86 °F)
  • Test principle: Colorimetric analysis


The test can be given to patients to check for an acute urinary tract infection, and to help monitor and assess therapy treatment.


Delivery information: Includes test strips, color chart, product insert.


Caution: The Rapid Response® urinary tract infection test strip is for diagnostic use and OTC in Canada and U.S.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR