VWR® DuraChill® Recirculating Chillers, -10 to 70 °C, 240 V
Supplier: VWR International
CA02A2P1-41AV1N CA02A2T1-41AV1N CA03A2P1-41AV1N CA03A2T1-41AV1N CA05A2P1-41AV1N CA05A2T1-41AV1N CA10A2P2-41AV1N CA10A2T1-41AV1N
76472-552EA 7106.81 USD
76472-552 76472-554 76472-560 76472-562 76472-568 76472-570 76472-576 76472-578
VWR® DuraChill® Recirculating Chillers, -10 to 70 °C, 240 V
Circulating Thermostats Closed Circulator Systems Cooling Circulators
DuraChill® chillers provide stable and reliable cooling for various common heat removal applications, including laser etching, AA furnaces, ICP, rotary evaporators, vacuum systems, reaction vessels, plasma etching, and condenser cooling.
  • Large, full-color touchscreen display with intuitive interface and support for five different languages
  • Compact, portable design takes up less floor space
  • DynamicFilter™ self-changing filter system
  • Front fill reservoir
  • Liquid level monitoring to prevent pump failure
  • Ultraviolet anti-biological light system
  • User-adjustable
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable
Electrical 240 V, 50 Hz