Environmental Express® Oakton® 250 and 260 Series Handheld Meters, Cole-Parmer
Supplier: Antyila Scientific
35660-26 35660-28 35660-30 35660-32 35660-34 35660-36 35660-38 35660-40 35660-42 35660-44 35660-46 35660-48 35660-50 35660-52
76471-934EA 1041.77 USD
76471-934 76471-936 76471-938 76471-940 76471-942 76471-944 76471-946 76471-948 76471-950 76471-952 76471-954 76471-956 76471-958 76471-960
Environmental Express® Oakton® 250 and 260 Series Handheld Meters, Cole-Parmer
Electrochemistry Meters pH and Multi-Parameter Meters
Reduce maintenance time with self-diagnostic function.
  • Rugged, waterproof, and dustproof with an IP 67 rating for use in any environment
  • Nonslip ergonomic design for comfortable handling
  • Easily view measurements with large monochrome LCD with white LED backlighting
  • Download and analyze data with USB output and phono jack printer connections
  • Take it with you—500 hours of battery life
Get easy-to-read and accurate readings with auto-hold, auto-stable, and real-time measurements with stability indicators. The Environmental Express® Oakton® 200 series ofMore Product Information