FectoVIR®-AAV Transfection Reagent for rAAV Production in Suspension, Polyplus-transfection
Supplier: Polyplus Transfection
101000044 101000022 101000004
76469-808EA 341.54 USD
76469-808 76469-810 76469-812
FectoVIR®-AAV Transfection Reagent for rAAV Production in Suspension, Polyplus-transfection
Transfection Reagents
Next generation of transfection reagent for rAAV production in suspension.
  • High performance: Superior yield for AAV production
  • Scalability: Developed for small to large scale manufacturing
  • Flexibility: Compatible with commercial culture media
  • Trustworthy: #1 transfection supplier of gene and cell therapy
  • GMP compliance: GMP grade coming soon
FectoVIR®-AAV is a novel class of animal-free transfection reagent specifically developed for research development up to industrial scale production of recombinant AAV (rAAV) viral vectors in suspension HEK-293 cell types.
FectoVIR®-AAV transfection reagent guarantees higher rAAV viral titers, improved flexibility and scalability for industrial scale manufacturing.
Certifications: Since 2002, Polyplus-transfection® is ISO 9001 certified and since then this certification has been successfully renewed every three years.
Order Now
SPECIFICATIONS
Cell Specific Suspension cells
Environmentally Preferable
Bioproduction Yes
DNA Yes
Virus production Yes