Corepoint Scientific™ Humidity and Temperature Stability Chambers, Horizon Scientific
Supplier: Horizon Scientific
NSRI231WSG/0H NSRI492WSG/0H NSRI231WSW/0H NSRI492WSW/0H
76469-444EA 33366.23 USD
76469-444 76469-446 76469-448 76469-450
Corepoint Scientific™ Humidity and Temperature Stability Chambers, Horizon Scientific
Testing Chambers Humidity Chambers
Outstanding accuracy and optimal performance on these models is achieved by a microprocessor-based PID controller which regulates user-settable temperature and humidity limits.
  • Proportional integral derivative (PID) controller with LCD display
  • Natural hydrocarbon refrigerant
  • Temperature setpoint range: 20 to 60 °C
  • Humidity range: 15 - 95% RH; humidity control: ±0.1% RH
These stability chambers are designed to provide controlled temperature or temperature and humidity testing environments. These chambers&More Product Information