VWR® Spinbar® Magnetic Stirring Bar, Cylindrical
Supplier: VWR International
76461-642
76461-642CS 1894.24 USD
76461-642
VWR® Spinbar® Magnetic Stirring Bar, Cylindrical
Stirrers Magnetic Stirring Bars
Cylindrical magnetic stirring bars offer excellent centering and smooth-running characteristics.
  • Small removable pivot ring in the center adds to bar versatility
  • Pivot ring minimizes the contact area of the bar with the vessel, reduces friction and lessens marring of plastic containers
  • Bar ends are slightly rounded
  • Alnico magnet encapsulated in FDA Grade, USP Class VI PTFE coating – superior chemical compatibility
  • Manufactured in a registered ISO 9001:2015 facility, as verified by SGS certification
  • Autoclave safe
Caution: TEFLON® is a registeredMore Product Information