SP Wilmad-LabGlass Thin Walled Quartz NMR Tubes, 5 mm, SP Industries
Supplier: SP Industries
535-PP-8QTZ-5 535-PP-9QTZ-5
14218-730PK 353.56 USD
14218-730 14218-736 14218-742 14218-966 14218-972 14218-978 14218-982 76459-866 76459-868
SP Wilmad-LabGlass Thin Walled Quartz NMR Tubes, 5 mm, SP Industries
Tubes NMR Tubes
Wilmad Precision NMR Quartz Tubes have an extremely low thermal expansion rate and high tensile strength which is 14 times more robust during the cooling/heating process than Type 1 Class A glass tubes.
  • Half the dielectric constant of Pyrex® glass helps improve the quality factor
  • Low Boron density at or below 0.1 ppm guarantees a clean background in Boron-11 NMR studies
  • Tight ID and OD tolerance as small as 0.0065 mm accommodates Wilmad inserts
  • To maximize SNR, Precision NMR Quartz Tubes have minimal paramagnetic impurities that would impact shimming
More Product Information